Assay Details
| Gene Symbol: gldc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348634.3 | NP_001335514.1 | 4 - 5 | 1218 | 64 | |
| NM_062808.8 | NP_495209.1 | 5 - 6 | 1518 | 64 | ||
More Information
Set Membership: |
|

gldc-1
Genomic Map