Assay Details
| Gene Symbol: rbmx-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062836.2 | NP_495237.1 | 2 - 3 | 337 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rbmx-2| Gene Symbol: rbmx-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062836.2 | NP_495237.1 | 2 - 3 | 337 | 108 | |
Set Membership: |
|