Assay Details
Gene Symbol: tat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383859.1 | 12 - 13 | 2763 | 133 | ||
NR_165322.1 | 12 - 13 | 2709 | 133 | |||
NR_165323.1 | 12 - 13 | 2709 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
