Assay Details
Gene Symbol: tat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383859.1 | 18 - 19 | 4445 | 67 | ||
NR_165322.1 | 18 - 19 | 4391 | 67 | |||
NR_165323.1 | 19 - 20 | 4512 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
