Assay Details
Gene Symbol: tat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383859.1 | 9 - 10 | 2241 | 132 | ||
NR_165322.1 | 9 - 10 | 2187 | 132 | |||
NR_165323.1 | 9 - 10 | 2187 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
