Assay Details
| Gene Symbol: F13H8.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062851.5 | NP_495252.1 | 7 - 8 | 1979 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F13H8.11| Gene Symbol: F13H8.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062851.5 | NP_495252.1 | 7 - 8 | 1979 | 86 | |
Set Membership: |
|