Assay Details
| Gene Symbol: F58F12.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062885.8 | NP_495286.1 | 2 - 3 | 261 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F58F12.1| Gene Symbol: F58F12.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062885.8 | NP_495286.1 | 2 - 3 | 261 | 70 | |
Set Membership: |
|