Assay Details
Gene Symbol: rsp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373731.2 | NP_001360098.1 | 4 - 5 | 690 | 59 | |
NM_062906.7 | NP_495307.3 | 3 - 4 | 534 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
