Assay Details
Gene Symbol: sra-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062928.2 | NP_495329.1 | 3 - 4 | 768 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
