Assay Details
| Gene Symbol: gld-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063078.4 | NP_495479.2 | 17 - 18 | 2471 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gld-3| Gene Symbol: gld-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063078.4 | NP_495479.2 | 17 - 18 | 2471 | 84 | |
Set Membership: |
|