Assay Details
| Gene Symbol: gld-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063078.4 | NP_495479.2 | 9 - 10 | 1019 | 95 | |
| NM_063079.7 | NP_495480.1 | 9 - 10 | 1019 | 95 | ||
More Information
Set Membership: |
|

gld-3
Genomic Map