Assay Details
| Gene Symbol: ada-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026962.3 | NP_001022133.1 | 5 - 6 | 1136 | 102 | |
| NM_001026963.5 | NP_001022134.1 | 3 - 4 | 847 | 102 | ||
More Information
Set Membership: |
|

ada-2
Genomic Map