Assay Details
Gene Symbol: ada-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026962.3 | NP_001022133.1 | 6 - 7 | 1365 | 95 | |
NM_001026963.5 | NP_001022134.1 | 4 - 5 | 1076 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
