Assay Details
Gene Symbol: insc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381495.1 | NP_001368642.1 | 6 - 7 | 642 | 71 | |
NM_001381496.2 | NP_001367460.1 | 6 - 7 | 666 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
