Assay Details
| Gene Symbol: syd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026975.4 | NP_001022146.1 | 12 - 13 | 1466 | 70 | |
| NM_001383891.1 | 10 - 11 | 1283 | 70 | |||
More Information
Set Membership: |
|

syd-1
Genomic Map