Assay Details
| Gene Symbol: syd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026975.4 | NP_001022146.1 | 5 - 6 | 498 | 93 | |
| NM_001383891.1 | 3 - 4 | 318 | 93 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383891.1 transcripts.
Set Membership: |
|

syd-1
Genomic Map