Assay Details
| Gene Symbol: sfxn-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026851.6 | NP_001022022.1 | 3 - 4 | 804 | 70 | |
| NM_001322747.3 | NP_001309542.1 | 2 - 3 | 509 | 70 | ||
More Information
Set Membership: |
|

sfxn-5
Genomic Map