Assay Details
| Gene Symbol: cct-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063349.8 | NP_495750.1 | 2 - 3 | 779 | 100 | |
| GenBank mRNA | U25697.1 | - | 2 - 3 | 779 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cct-4| Gene Symbol: cct-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063349.8 | NP_495750.1 | 2 - 3 | 779 | 100 | |
| GenBank mRNA | U25697.1 | - | 2 - 3 | 779 | 100 | |
Set Membership: |
|