Assay Details
Gene Symbol: F54C9.9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063414.5 | NP_495815.1 | 5 - 6 | 775 | 161 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
