Assay Details
Gene Symbol: cutl-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322749.3 | NP_001309578.1 | 6 | 827 | 148 | |
NM_001322750.3 | NP_001309579.1 | 3 - 4 | 409 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
