Assay Details
| Gene Symbol: cutl-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322749.3 | NP_001309578.1 | 7 | 1001 | 146 | |
| NM_001322750.3 | NP_001309579.1 | 4 - 5 | 583 | 146 | ||
More Information
Set Membership: |
|

cutl-12
Genomic Map