Assay Details
| Gene Symbol: rha-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373779.1 | NP_001360473.1 | 10 - 11 | 2899 | 110 | |
| NM_063489.6 | NP_495890.2 | 10 - 11 | 2899 | 110 | ||
More Information
Set Membership: |
|

rha-1
Genomic Map