Assay Details
Gene Symbol: rha-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373779.1 | NP_001360473.1 | 1 - 2 | 173 | 112 | |
NM_063489.6 | NP_495890.2 | 1 - 2 | 173 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
