Assay Details
| Gene Symbol: cal-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063505.5 | NP_495906.2 | 1 - 2 | 158 | 98 | |
| GenBank mRNA | AJ512489.1 | - | 1 - 2 | 138 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cal-2| Gene Symbol: cal-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063505.5 | NP_495906.2 | 1 - 2 | 158 | 98 | |
| GenBank mRNA | AJ512489.1 | - | 1 - 2 | 138 | 98 | |
Set Membership: |
|