Assay Details
| Gene Symbol: dep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267263.3 | NP_001254192.1 | 3 - 4 | 937 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dep-1| Gene Symbol: dep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267263.3 | NP_001254192.1 | 3 - 4 | 937 | 101 | |
Set Membership: |
|