Assay Details
| Gene Symbol: chmp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063532.8 | NP_495933.1 | 2 - 3 | 257 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chmp-7| Gene Symbol: chmp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063532.8 | NP_495933.1 | 2 - 3 | 257 | 91 | |
Set Membership: |
|