Assay Details
| Gene Symbol: dpl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063556.8 | NP_495957.1 | 7 - 8 | 1140 | 109 | |
| GenBank mRNA | AY028165.1 | - | 7 - 8 | 1127 | 109 | |
More Information
Set Membership: |
|

dpl-1
Genomic Map