Assay Details
Gene Symbol: kin-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063616.6 | NP_496017.1 | 3 - 4 | 549 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
