Assay Details
| Gene Symbol: glt-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063693.5 | NP_496094.1 | 6 - 7 | 1278 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

glt-5| Gene Symbol: glt-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063693.5 | NP_496094.1 | 6 - 7 | 1278 | 86 | |
Set Membership: |
|