Assay Details
| Gene Symbol: lov-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063783.4 | NP_496184.3 | 28 - 29 | 8413 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lov-1| Gene Symbol: lov-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063783.4 | NP_496184.3 | 28 - 29 | 8413 | 94 | |
Set Membership: |
|