Assay Details
Gene Symbol: pax-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063788.3 | NP_496189.2 | 5 - 6 | 773 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
