Assay Details
Gene Symbol: srb-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063798.1 | NP_496199.1 | 4 - 5 | 531 | 191 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
