Assay Details
| Gene Symbol: plc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063804.5 | NP_496205.2 | 12 - 13 | 3957 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plc-3| Gene Symbol: plc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063804.5 | NP_496205.2 | 12 - 13 | 3957 | 71 | |
Set Membership: |
|