Assay Details
| Gene Symbol: gcy-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063817.2 | NP_496218.2 | 8 - 9 | 1320 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-4| Gene Symbol: gcy-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063817.2 | NP_496218.2 | 8 - 9 | 1320 | 124 | |
Set Membership: |
|