Assay Details
| Gene Symbol: stip-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373788.1 | NP_001359529.1 | 2 - 3 | 186 | 79 | |
| NM_063825.6 | NP_496226.2 | 2 - 3 | 174 | 79 | ||
| GenBank mRNA | DQ342049.1 | - | 2 - 3 | 196 | 79 | |
More Information
Set Membership: |
|

stip-1
Genomic Map