Assay Details
Gene Symbol: dap-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063879.6 | NP_496280.1 | 1 - 2 | 215 | 70 | |
GenBank mRNA | AF058329.1 | - | 1 - 2 | 253 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
