Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 15 - 16 | 1638 | 75 | |
| NM_001026868.5 | NP_001022039.2 | 15 - 16 | 1619 | 75 | ||
| NM_001083157.4 | NP_001076626.2 | 9 - 10 | 1031 | 75 | ||
| NM_001129127.4 | NP_001122599.1 | 8 - 9 | 977 | 75 | ||
| NM_001129129.4 | NP_001122601.1 | 13 - 14 | 1460 | 75 | ||
| NM_001377853.2 | NP_001364655.1 | 16 - 17 | 1691 | 75 | ||
| NM_001377854.2 | NP_001364657.1 | 15 - 16 | 1604 | 75 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map