Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 19 - 20 | 2670 | 87 | |
| NM_001083157.4 | NP_001076626.2 | 13 - 14 | 2063 | 87 | ||
| NM_001129127.4 | NP_001122599.1 | 12 - 13 | 2009 | 87 | ||
| NM_001129129.4 | NP_001122601.1 | 17 - 18 | 2492 | 87 | ||
| NM_001377853.2 | NP_001364655.1 | 20 - 21 | 2723 | 87 | ||
| NM_001377854.2 | NP_001364657.1 | 19 - 20 | 2636 | 87 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map