Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 30 - 31 | 3767 | 76 | |
| NM_001083157.4 | NP_001076626.2 | 24 - 25 | 3160 | 76 | ||
| NM_001129127.4 | NP_001122599.1 | 23 - 24 | 3106 | 76 | ||
| NM_001129129.4 | NP_001122601.1 | 28 - 29 | 3589 | 76 | ||
| NM_001267333.3 | NP_001254262.1 | 7 - 8 | 781 | 76 | ||
| NM_001377853.2 | NP_001364655.1 | 31 - 32 | 3820 | 76 | ||
| NM_001377854.2 | NP_001364657.1 | 30 - 31 | 3733 | 76 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map