Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 31 - 32 | 4332 | 71 | |
| NM_001083157.4 | NP_001076626.2 | 25 - 26 | 3725 | 71 | ||
| NM_001129127.4 | NP_001122599.1 | 24 - 25 | 3671 | 71 | ||
| NM_001129129.4 | NP_001122601.1 | 29 - 30 | 4154 | 71 | ||
| NM_001267333.3 | NP_001254262.1 | 8 - 9 | 1346 | 71 | ||
| NM_001267334.3 | NP_001254263.1 | 1 - 2 | 59 | 71 | ||
| NM_001377853.2 | NP_001364655.1 | 32 - 33 | 4385 | 71 | ||
| NM_001377854.2 | NP_001364657.1 | 31 - 32 | 4298 | 71 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map