Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 4 - 5 | 307 | 82 | |
| NM_001026868.5 | NP_001022039.2 | 4 - 5 | 288 | 82 | ||
| NM_001129129.4 | NP_001122601.1 | 2 - 3 | 129 | 82 | ||
| NM_001377853.2 | NP_001364655.1 | 5 - 6 | 360 | 82 | ||
| NM_001377854.2 | NP_001364657.1 | 4 - 5 | 273 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001129129.4 transcripts.
Set Membership: |
|

mpz-1
Genomic Map