Assay Details
| Gene Symbol: ect-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063917.6 | NP_496318.1 | 6 - 7 | 854 | 101 | |
| NM_063918.3 | NP_496319.1 | 7 - 8 | 879 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ect-2| Gene Symbol: ect-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063917.6 | NP_496318.1 | 6 - 7 | 854 | 101 | |
| NM_063918.3 | NP_496319.1 | 7 - 8 | 879 | 101 | ||
Set Membership: |
|