Assay Details
| Gene Symbol: sra-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026991.4 | NP_001022162.1 | 1 - 2 | 249 | 135 | |
| NM_001026992.3 | NP_001022163.1 | 1 - 2 | 257 | 135 | ||
More Information
Set Membership: |
|

sra-10
Genomic Map