Assay Details
| Gene Symbol: mat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063982.4 | NP_496383.2 | 3 - 4 | 1176 | 156 | |
| Gene Symbol: apc-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063980.3 | NP_496381.1 | 6 - 7 | 2177 | 156 | |
More Information
Set Membership: |
|

mat-2
Genomic Map