Assay Details
| Gene Symbol: aars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064043.7 | NP_496444.1 | 8 - 9 | 2788 | 68 | |
| NR_023953.1 | 8 - 9 | 2286 | 68 | |||
| GenBank mRNA | AF188715.1 | - | 8 - 9 | 2367 | 68 | |
More Information
Set Membership: |
|

aars-1
Genomic Map