Assay Details
| Gene Symbol: nadk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064164.7 | NP_496565.2 | 1 - 2 | 65 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_064164.7 transcripts.
Set Membership: |
|

nadk-2
Genomic Map