Assay Details
| Gene Symbol: nadk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064164.7 | NP_496565.2 | 5 - 6 | 744 | 94 | |
| NM_064165.7 | NP_496566.1 | 5 - 6 | 875 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nadk-2| Gene Symbol: nadk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064164.7 | NP_496565.2 | 5 - 6 | 744 | 94 | |
| NM_064165.7 | NP_496566.1 | 5 - 6 | 875 | 94 | ||
Set Membership: |
|