Assay Details
| Gene Symbol: nadk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064164.7 | NP_496565.2 | 7 - 8 | 1289 | 86 | |
| NM_064165.7 | NP_496566.1 | 7 - 8 | 1420 | 86 | ||
More Information
Set Membership: |
|

nadk-2
Genomic Map