Assay Details
Gene Symbol: ras-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064222.7 | NP_496623.1 | 4 - 5 | 508 | 101 | |
GenBank mRNA | U56930.1 | - | 4 - 5 | 483 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
