Assay Details
| Gene Symbol: cnp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064314.6 | NP_496715.3 | 14 - 15 | 1747 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cnp-2| Gene Symbol: cnp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064314.6 | NP_496715.3 | 14 - 15 | 1747 | 112 | |
Set Membership: |
|